missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1019952500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1019952500 2.5 L Glass Bottle
M1019951000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1019952500 Supplier Technidata Supplier No. 1.01995.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -108.6°C
Color Colorless
Density 0.74g/cm3 at 20°C
Quantity 2.5 L
Flash Point −28°C (−18.4°F)
Ignition Point 460°C (860°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 268 hPa (20°C)
UN Number 2398
Boiling Point 55.3°C
Solubility Information Soluble in water: 42g/L at 20°C
Formula Weight 88.15
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H315 Causes skin irritation.
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.