missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydroperoxide, 70% aq. soln.

Catalog No. AAA13926AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13926AP 500 mL
AAA13926AE 100 mL
AAA139260F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13926AP Supplier Thermo Scientific Chemicals Supplier No. A13926AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-91-2 | C4H10O2 | 90.122 g/mol

tert-Butyl hydroperoxide is used as an initiator for radical polymerization and in various oxidation process such as sharpless epoxidation. It is involved in osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions. Furthermore, it is used in catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliary and in the oxidation of dibenzothiophenes. It plays an important role for the introduction of peroxy groups in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydroperoxide is used as an initiator for radical polymerization and in various oxidation process such as sharpless epoxidation. It is involved in osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions. Furthermore, it is used in catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliary and in the oxidation of dibenzothiophenes. It plays an important role for the introduction of peroxy groups in organic synthesis.

Solubility
Miscible with water and diethyl ether.

Notes
Light sensitive. Store in cool place. Incompatible with powdered metals, strong oxidizing agents, reducing agents, acids, alkalis and heavy metals.

Chemical Identifiers

CAS 75-91-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00002130
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
Synonym tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
PubChem CID 6410
ChEBI CHEBI:64090
IUPAC Name 2-hydroperoxy-2-methylpropane
SMILES CC(C)(C)OO

Specifications

Melting Point °C to 3°C
Density 0.937
Boiling Point 96°C (decomposition)
Flash Point 43°C (109°F)
Assay Percent Range 70% Aqueous Solution
Refractive Index 1.387
Linear Formula (CH3)3COOH
Quantity 500 mL
UN Number UN3109
Beilstein 1098280
Merck Index 14,1570
Solubility Information Miscible with water and diethyl ether.
Formula Weight 90.12
Chemical Name or Material tert-Butyl Hydroperoxide
Show More Show Less
Hazard Category H226-H242-H302-H311-H314-H317-H330-H335-H341
Hazard Statement GHS H Statement
H300-H310-H330-H314-H226-H242
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Heating may cause a fire.
Precautionary Statement P201-P202-P210-P220-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P37
DOTInformation Hazard Class: 5.2
EINECSNumber 200-915-7
RTECSNumber EQ4900000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only