missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Ethyl Ether 95.0+%, TCI America™

Catalog No. B15425ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15425ML 5 mL
B154225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15425ML Supplier TCI America Supplier No. B15425ML

Chemical Identifiers

CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C

Specifications

Color Colorless
Boiling Point 70°C
Quantity 5 mL
UN Number 1179
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Ethyl Ether
EINECSNumber (2)-4058
RTECSNumber KN4730200
TSCA Yes