missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Bromoacetate 98.0+%, TCI America™

Catalog No. B147325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B147325G 25 g
B1473250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B147325G Supplier TCI America Supplier No. B147325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5292-43-3
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000188
InChI Key BNWCETAHAJSBFG-UHFFFAOYSA-N
Synonym tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
PubChem CID 79177
IUPAC Name tert-butyl 2-bromoacetate
SMILES CC(C)(C)OC(=O)CBr

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 g
UN Number 2924
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Bromoacetate