missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 361345-40-6
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
InChI Key VKSINZNBWVJILC-UHFFFAOYSA-N
Synonym tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
PubChem CID 7127799
IUPAC Name tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC30414CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30414DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30414DE
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

361345-40-6
291.395
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
C16H25N3O2
VKSINZNBWVJILC-UHFFFAOYSA-N
7127799
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

361345-40-6
C16H25N3O2
VKSINZNBWVJILC-UHFFFAOYSA-N
tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
7127799
97%
97%
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N
291.395
291.39
tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
Videos
SDS
Documents

Documents