missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC30414DE
| Quantity | 5g |
|---|
Chemical Identifiers
| 361345-40-6 | |
| 291.395 | |
| tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected | |
| tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate |
| C16H25N3O2 | |
| VKSINZNBWVJILC-UHFFFAOYSA-N | |
| 7127799 | |
| CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N |
Specifications
| tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate | |
| 97% | |
| C16H25N3O2 | |
| tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected | |
| CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N | |
| 291.395 | |
| 291.39 |
| 361345-40-6 | |
| Amber Glass Bottle | |
| 5g | |
| VKSINZNBWVJILC-UHFFFAOYSA-N | |
| tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate | |
| 7127799 | |
| 97% |