missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC30414DA

Quantity 1g

 View more versions of this product

Catalog No. CC30414DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

361345-40-6
291.395
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
C16H25N3O2
VKSINZNBWVJILC-UHFFFAOYSA-N
7127799
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
97%
C16H25N3O2
tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N
291.395
291.39
361345-40-6
Amber Glass Bottle
1g
VKSINZNBWVJILC-UHFFFAOYSA-N
tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
7127799
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions