missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-(Isopropyl)carbazate 98.0+%, TCI America™

Catalog No. B43755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43755G 5 g
B43751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43755G Supplier TCI America Supplier No. B43755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16689-35-3
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD14635748
InChI Key PUAKAEDMCXRDPR-UHFFFAOYSA-N
Synonym 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 1-Boc-2-isopropylhydrazine
PubChem CID 11217534
IUPAC Name N'-(propan-2-yl)(tert-butoxy)carbohydrazide
SMILES CC(C)NNC(=O)OC(C)(C)C

Specifications

Melting Point 54°C
Color White
Boiling Point 80°C
Quantity 5 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 3-(Isopropyl)carbazate