missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Terrific Broth
GREENER_CHOICE

Catalog No. A1374301
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1374301 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1374301 Supplier Gibco™ Supplier No. A1374301

Gibco Terrific Broth is a ready-to-use liquid growth medium used for bacterial culture. Our filter-sterilized Terrific Broth can be used for nearly all research applications that require a liquid bacterial culturing step, such as protein expression, cloning, and expansion.

Gibco Terrific Broth is a ready-to-use liquid growth medium used for bacterial culture. Our filter-sterilized Terrific Broth can be used for nearly all research applications that require a liquid bacterial culturing step, such as protein expression, cloning, and expansion.

Terrific Broth is a nutritionally rich bacterial medium for higher density growth and maintenance of recombinant E. coli strains. It was specifically developed to achieve higher plasmid DNA yield from E. coli and can yield 4–7 times more DNA from pUC-based vectors compared to LB medium. This versitle broth is also useful for processing a large number of different clones from smaller cultures. 

Terrific Broth formulation per liter
11.8 g Tryptone
23.6 g Yeast extract
9.4 g Dipotassium hydrogen phosphate
2.2 g Potassium dihydrogen phosphate
4 mL Glycerol (0.4%)

• TB medium contains higher concentration of yeast extract compared to other media
• Offers better buffering capacity due higher salt concentrations
• Added glycerol has a stabilizing effect on recombinant proteins

Primary applications
• Growth of E. coli strains to achieve higher plasmid yield
• Expression of recombinant proteins using bacteria

Gibco media bottles
Each media bottle features an angled opening for simplified pipetting. The 1 L Gibco media bottle uses 39% less source material than comparable media bottles on the market. Both the cap and bottle can be recycled as they are made from highly recyclable high-density polyethylene (HDPE) and polyethylene terephthalate (PETE).

Find the media format that fits your needs
We offer a variety of ready-to-use media formulations, such as LB Broth (Cat. No. 10855021) or S.O.C recovery medium (Cat. No. 15544034).

Specifications

Content And Storage • 1 L Terrific Broth Liquid

Store at room temperature.

Media Type TB
Preparation Method Ready-to-Use
Product Type Liquid Medium
Sterility Sterile
Target Organism Class Coliforms
Quantity 1 L
Shipping Condition Room Temperature
Species Escherichia coli
How can unstable or toxic DNA inserts be maintained in bacteria?

There are a few steps you can take to improve stability of clones with difficult-to-maintain inserts. Supplement the medium with extra nutrients (e.g., add 20-30 mM glucose to Terrific Broth) or try a vector that has a reduced copy number (e.g., pBR322). Some clones can exhibit a high degree of deletions; this is usually a result of the clones having long terminal repeat (LTR) sequences or regions with high secondary structure. To overcome this problem, the cells can be grown at 30°C or ambient temperature (in LB or in a nutrient rich broth like Terrific Broth). Do not to let the cells reach late stationary phase in liquid culture. Alternatively, transform into cells that maintain unstable sequences such as Stbl2, Stbl3, or Stbl4 cells.

Do you offer an alternative to Terrific Broth (Cat. No. A1374301)?

We offer Terrific Broth (Cat. No. 22711022) as an alternative to Terrific Broth (Cat. No. A1374301).

For Research Use Only. Not for use in diagnostic procedures.