missing translation for 'onlineSavingsMsg'
Learn More

α-Terpineol, ≥90.0% (GC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111016004
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111016004 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111016004 Fournisseur Merck Emd Millipore Code fournisseur 30627100MG
Il en reste null
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Spécifications

CAS 98-55-5
Density 0.93 g/mL (at 25°C)
Flash Point 90°C - Closed Cup
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00001557
Melting Point 31°C to 35°C
Boiling Point 217°C to 218°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 154.25
Percent Purity ≥90.0% (GC)
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats