Learn More
LGC Genomics Llc (Lucigen) Terminator 5'-Phosphate-Dependent Exonuclease, 40U @ 1 U/LA processive 5 to 3 riboexonuclease that specifically digests RNA with 5-monophosphate ends only.
Numéro de catalogue. NC1460234
Encompass
Terminator™ 5-Phosphate-Dependent Exonuclease is a processive 53 exonuclease that digests RNA that has a 5-monophosphate end. It does not digest RNA that has a 5-triphosphate, 5-cap or 5-hydroxyl group.
Numéro de catalogue. NC1460234 Fournisseur LGC Genomics Llc Code fournisseur TER51020
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Terminator™ 5-Phosphate-Dependent Exonuclease is a processive 53 exonuclease that digests RNA that has a 5-monophosphate end. It does not digest RNA that has a 5-triphosphate, 5-cap or 5-hydroxyl group.