missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Teriflunomide

Catalog No. AC467112500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467112500 250 mg
AC467110010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467112500 Supplier Thermo Scientific Chemicals Supplier No. 467112500
Only null left

CAS: 163451-81-8 | C12H9F3N2O2, C12H9F3N2O2 | 270.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163451-81-8
Molecular Formula C12H9F3N2O2

Specifications

Quantity 250 mg
Melting Point 229.0°C
Formula Weight 270.21
InChI Key UTNUDOFZCWSZMS-UHFFFAOYSA-N
SMILES CC(O)=C(C#N)C(=O)NC1=CC=C(C=C1)C(F)(F)F
IUPAC Name 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide
Molecular Weight (g/mol) 270.21
Chemical Name or Material Teriflunomide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only