missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TER-119 Monoclonal Antibody (TER-119), eFluor™ 450, eBioscience™, Invitrogen™

Numéro de catalogue. 5016381
Modifier l'affichage
Click to view available options
Quantité:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
5016381 25 μg
5016382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 5016381 Fournisseur Invitrogen™ Code fournisseur 48592180
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rat Monoclonal Antibody

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin. Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.

Spécifications

Antigène TER-119
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 0.2 mg/mL
Conjugué eFluor 450
Formule PBS with 0.09% sodium azide; pH 7.2
Gène Ly76
Numéro d’acquisition du gène 0
Alias du gène Ly76; Ly-76; TER119
Symboles de gène Ly76
Espèces hôtes Rat
Méthode de purification Affinity chromatography
Quantité 25 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 104231
Espèces cibles Mouse
Contenu et stockage 4°C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgG2b κ
Afficher plus de résultats Afficher moins de résultats

For Research Use Only.