missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEPP Polyclonal Antibody

Description
Immunogen sequence: DRDTVKACLPD EHCQSTTYCR KDEFDNAHFT LLGVPNKPLQ CLDITATGQK LRNRYHEGKL APIAPGINRV DW Highest antigen sequence identity to the following orthologs - mouse 77%, rat 70%.
TEPP is a protein coding gene.
Spécifications
Spécifications
| Antigen | TEPP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TEPP |
| Gene Accession No. | Q6URK8 |
| Gene Alias | TEPP; testicular tissue protein Li 196; testis, prostate and placenta expressed; testis, prostate and placenta-expressed protein; testis/prostate/placenta-expressed protein |
| Gene Symbols | TEPP |
| Afficher plus de résultats |