missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tenofovir

Catalog No. AC465610050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465610050 5 g
AC465610010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465610050 Supplier Thermo Scientific Chemicals Supplier No. 465610050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 147127-20-6 | C9H14N5O4P, C9 H14 N5 O4 P | 287.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147127-20-6
Molecular Formula C9 H14 N5 O4 P

Specifications

Quantity 5 g
Formula Weight 287.21
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
Chemical Name or Material Tenofovir

RUO – Research Use Only