accessibility menu, dialog, popup

UserName

CAS RN 147127-20-6

IUPAC Name: ({[(2R)-1-(6-amino-9H-purin-9-yl)propan-2-yl]oxy}methyl)phosphonic acid
Synonyms: tenofovir anhydrous tenofovir pmpa gel apropovir tenofovir (anhydrous) tenofovir (anh.) TDF PMPA anh. tenofovir microbicides for sexually transmitted diseases
Molecular Weight (g/mol): 287.22
Molecular Formula: C9H14N5O4P
InChi Key: SGOIRFVFHAKUTI-ZCFIWIBFSA-N
SMILES: C[C@H](CN1C=NC2=C(N)N=CN=C12)OCP(O)(O)=O
Physical Form 
  • (2)
  • (2)
Product Type 
  • (2)
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)