missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) ethoxide, tech. 85%, remainder ethanol

Catalog No. AA4110509
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4110509 10 g
AA4110504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4110509 Supplier Thermo Scientific Chemicals Supplier No. 04110509

CAS: 2017-01-8 | C6H14N2O | 130.19 g/mol

Tellurium(IV) ethoxide could be used in sol-gel processing for the fabrication of tellurite thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2017-01-8
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00270937
InChI Key RWIVICVCHVMHMU-UHFFFAOYSA-N
PubChem CID 408285
IUPAC Name 2-morpholin-4-ylethanamine
SMILES C1COCCN1CCN

Specifications

Melting Point >100°C (decomposition)
Density 1.52 g/mL
Boiling Point 88°C to 90°C (2 mmHg)
Flash Point 27°C (81°F)
Odor Alcohol-like
Quantity 10 g
UN Number UN1993
Sensitivity Light and moisture sensitive
Synonym 4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
Solubility Information Decomposes in water to TeO2
Formula Weight 307.84
Percent Purity 85%
Grade Technical
Physical Form Liquid
Assay Remainder ethanol
Chemical Name or Material Tellurium(IV) ethoxide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only