Learn More
MilliporeSigma™ Supelco™ Tedlar™ SCV Gas Sampling Bag with Thermogreen™ LB-2 Septa
Numéro de catalogue. 111009536
Change view
Click to view available options
Capacity (Metric):
0.6 L
1 L
2 L
3 L
5 L
10 L
25 L
Quantity:
10/Pk.
5/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric) Quantity
111009536 10 L 10/Pk.
111010525 0.6 L 10/Pk.
111016273 1 L 10/Pk.
111010237 2 L 10/Pk.
111010002 3 L 10/Pk.
111009858 5 L 10/Pk.
111009493 25 L 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Numéro de catalogue. 111009536 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 30276U
Il en reste null
Tedlar™ is recommended in several US EPA methods for sampling VOCs and sulfur compounds.
Tedlar™ is made from a high-quality 2 mil Tedlar™ film for superior inertness, impermeability, sample integrity and good recovery data.
Spécifications
Includes Screw Cap Valve (SCV)
Material PVDF Film
Product Line Tedlar
Quantity 10/Pk.
Capacity (Metric) 10 L
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company. Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC.