missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tedizolid

Catalog No. AC467582500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467582500 250 mg
AC467580010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467582500 Supplier Thermo Scientific Chemicals Supplier No. 467582500
Only null left

CAS: 856866-72-3 | C17H15FN6O3, C17H15FN6O3 | 370.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 856866-72-3
Molecular Formula C17H15FN6O3

Specifications

Quantity 250 mg
Melting Point 201.0°C
Formula Weight 370.34
InChI Key XFALPSLJIHVRKE-UHFFFAOYNA-N
SMILES CN1N=NC(=N1)C1=NC=C(C=C1)C1=C(F)C=C(C=C1)N1CC(CO)OC1=O
IUPAC Name 3-{3-fluoro-4-[6-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 370.34
Chemical Name or Material Tedizolid

RUO – Research Use Only