SignalChem Biotech PI3K (p110 delta (E1021K)/p85 alpha), Active 500ug
Numéro de catalogue. 502573770
Recombinant full-length human p110 delta (E1021K) and human p85 alpha were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
Numéro de catalogue.
502573770
Fournisseur
SignalChem BiotechCode fournisseur
P30122H500
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant full-length human p110 delta (E1021K) and human p85 alpha were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.