missing translation for 'onlineSavingsMsg'
Learn More

Tebuthiuron 98.0+%, TCI America™

Numéro de catalogue. T36475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T36475G 5 g
T364725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T36475G Fournisseur TCI America Code fournisseur T36475G
Il en reste null

Identifiants chimiques

CAS 34014-18-1
Molecular Formula C9H16N4OS
Molecular Weight (g/mol) 228.314
MDL Number MFCD00078732
InChI Key HBPDKDSFLXWOAE-UHFFFAOYSA-N
Synonym tebuthiuron, spike, graslan, perflan, brulan, tiurolan, prefmid, tebulan, brush bullet, 1-5-tert-butyl-1,3,4-thiadiazol-2-yl-1,3-dimethylurea
PubChem CID 5383
ChEBI CHEBI:81745
IUPAC Name 1-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-1,3-dimethylurea
SMILES CC(C)(C)C1=NN=C(S1)N(C)C(=O)NC

Spécifications

Melting Point 163°C
Color White-Yellow
pH 5 - 9
Quantity 5 g
UN Number 3077
Formula Weight 228.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tebuthiuron
EINECSNumber (5)-5242
RTECSNumber YS4250000
TSCA No