missing translation for 'onlineSavingsMsg'
Learn More

TE buffer, pH 8.0, RNAse free

Catalog No. AAJ62745EQE
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62745EQE 125 mL
AAJ62745AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62745EQE Supplier Thermo Scientific Chemicals Supplier No. J62745EQE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Tris-EDTA (TE) buffer solution, pH 8.0 may also be used as a washing buffer. TE buffer is often used to store DNA and RNA while protecting it from degradation. Some protocols use TE 10:0.1 with 0.1 mM EDTA to reduce the interaction of the EDTA with downstream applications.

Solubility
Not miscible or difficult to mix in water.

Notes
200mM Tris-HCl with 20mM EDTA

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

pH 8.0
Physical Form Liquid
Quantity 125 mL
Solubility Information Not miscible or difficult to mix in water.
Chemical Name or Material TE buffer, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only