missing translation for 'onlineSavingsMsg'
Learn More

TE buffer, pH 7.4, RNAse free

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

$132.84 - $233.29

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60234EQE
View Documents
Thermo Scientific Chemicals
J60234EQE
125 mL
Each for $132.84
Only null left
 
AAJ60234AK
View Documents
Thermo Scientific Chemicals
J60234AK
250 mL
Each for $233.29
Only null left
 
Description

Description

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

Solubility
Fully miscible in water.

Notes
200mM Tris-HCl with 20mM EDTA
Specifications

Chemical Identifiers

1185-53-1
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C4H12ClNO3
MFCD00236359
93573
C(C(CO)(CO)N)O.Cl

Specifications

1185-53-1
7.4
125 mL
MFCD00236359
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Undesignated
Liquid
C4H12ClNO3
Fully miscible in water.
C(C(CO)(CO)N)O.Cl
157.594
TE buffer, RNAse free
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only