missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TE buffer, (20X), pH 8.0, autoclaved

Catalog No. AAJ62388AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62388AP 500 mL
AAJ62388AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62388AP Supplier Thermo Scientific Chemicals Supplier No. J62388AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids. | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Physical Form Liquid
Quantity 500 mL
Packaging Plastic bottle
Solubility Information Fully miscible in water.
Chemical Name or Material TE buffer
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only