Learn More
TE buffer, (20X, pH 7.6), RNAse free
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
$134.68 - $134.68
Chemical Identifiers
| CAS | 1185-53-1 |
|---|---|
| Molecular Formula | C4H12ClNO3 |
| Molecular Weight (g/mol) | 157.594 |
| MDL Number | MFCD00236359 |
| InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
| PubChem CID | 93573 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| SMILES | C(C(CO)(CO)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62285EQE
|
Thermo Scientific Chemicals
J62285EQE |
125 mL |
Each for $134.68
|
|
|||||
Description
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. Â TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTE buffer is a commonly used buffer solution in procedures involving DNA or RNA. Â TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.
Solubility
Fully miscible in water.
Notes
200mM Tris-HCl with 20mM EDTA
Chemical Identifiers
| 1185-53-1 | |
| 157.594 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| C4H12ClNO3 | |
| MFCD00236359 | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
Specifications
| 1185-53-1 | |
| Liquid | |
| C4H12ClNO3 | |
| Fully miscible in water. | |
| C(C(CO)(CO)N)O.Cl | |
| 157.594 | |
| TE buffer, RNAse free |
| 7.6 | |
| 125 mL | |
| MFCD00236359 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only