missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure

Catalog No. AAJ75893XCR
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
Packaging:
Nalgene bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ75893XCR 10 x 1 mL Nalgene bottle
AAJ75893AE 100 mL Nalgene bottle
AAJ75893AP 500 mL Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ75893XCR Supplier Thermo Fisher Scientific Supplier No. J75893XCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use directly to resuspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Use directly to resuspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Physical Form Solution
Quantity 10 x 1 mL
Packaging Nalgene bottle
Grade Molecular Biology
Chemical Name or Material TE Buffer
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only