missing translation for 'onlineSavingsMsg'
Learn More

TCPP 97.0+%, TCI America™

Catalog No. A50151G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50151G 1 g
A5015100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50151G Supplier TCI America Supplier No. A50151G

[For the simultaneous determination of metals by HPLC]

Chemical Identifiers

CAS 14609-54-2
Molecular Formula C48H30N4O8
Molecular Weight (g/mol) 790.788
MDL Number MFCD00064860
InChI Key SMOZAZLNDSFWAB-UHFFFAOYSA-N
Synonym Tetrakis(4-carboxyphenyl)porphyrin
PubChem CID 5479495
IUPAC Name 4-[10,15,20-tris(4-carboxyphenyl)-21,24-dihydroporphyrin-5-yl]benzoic acid
SMILES C1=CC(=CC=C1C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)C(=O)O)C8=CC=C(C=C8)C(=O)O)C9=CC=C(C=C9)C(=O)O)N3)C(=O)O

Specifications

Color Purple
Quantity 1 g
Formula Weight 790.79
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material TCPP [=Tetrakis(4-carboxyphenyl)porphyrin] [Ultra-high sensitive spectrophotometric reagent f
RTECSNumber DH6488300
TSCA No