missing translation for 'onlineSavingsMsg'
Learn More

TCLP Extraction Fluid No. 1, Certified, LabChem™

Catalog No. LC259956
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass
Cubitainer™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC259956 10 L Cubitainer™
LC259954 4 L Amber Glass
LC259955 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC259956 Supplier LabChem Supplier No. LC259956

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material TCLP Extraction Fluid No. 1
Packaging Cubitainer™
CAS Max % 99.18, 0.57, 0.25
CAS 64-19-7, 1310-73-2
Identification Passes Test
Viscosity 1.02cSt
Density 0.999g/mL
Physical Form Liquid
Linear Formula CH3COOH
Color Colorless
pH 4.88 to 4.98
Solubility Soluble in water
Quantity 10 L
Formula Weight 60.05
Grade Certified
Show More Show Less