missing translation for 'onlineSavingsMsg'
Learn More

TCL Volatiles Mix, Certified Reference Material, 2,000 μg/mL Each Component in Methanol: Water (9:1), MilliporeSigma™ Supelco™

Catalog No. 111017306
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017306 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017306 Supplier Merck Emd Millipore Supplier No. CRM48949

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 2,000 μg/mL each component in methanol: water (9:1)
Synonym EPA TCL Volatiles Mix 1
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Acetone; 2-Butanone; 2-Hexanone; 4-Methyl-2-pentanone

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC