missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557816
Description
Immunogen sequence: SEGSSQPEEQ LKSPMAESLD GGCSSSSEDQ GERVRQLSGQ STSSDTTYKG GASEKAGSSP AQGAQNEPPR LNASPAAREE ATSPGAKDMP LSSDGNPKVN EKTVGVIVS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
This gene encodes a transcription factor that recognizes the platelet-derived growth factor-responsive element in the matrix metalloproteinase 3 promoter. The encoded protein is thought to be a transcriptional coactivator, enhancing the activity of transcription factors such as JUN and SP1. Mutations in this gene are associated with autism spectrum disorders. Alternative splicing results in multiple transcript variants.Identifiants chimiques
| TCF20 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| TCF20 | |
| 2810438H08Rik; AR1; KIAA0292; mKIAA0292; Nuclear factor SPBP; Protein AR1; SPBP; SPRE-binding protein; stromelysin 1 PDGF responsive element binding protein; stromelysin 1 PDGF-responsive element-binding protein; stromelysin PDGF responsive element binding protein transcription factor; stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; Tcf20; TCF-20; Transcription factor 20; transcription factor 20 (AR1) | |
| TCF20 | |
| Primary | |
| Antigen Affinity Chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGU0 | |
| 6942 | |
| Recombinant protein corresponding to Human TCF20. Recombinant protein control fragment (Product #RP-95352). | |
| Antibody |
Spécifications
| TCF20 | |
| Polyclonal | |
| Unconjugated | |
| TCF20 | |
| 2810438H08Rik; AR1; KIAA0292; mKIAA0292; Nuclear factor SPBP; Protein AR1; SPBP; SPRE-binding protein; stromelysin 1 PDGF responsive element binding protein; stromelysin 1 PDGF-responsive element-binding protein; stromelysin PDGF responsive element binding protein transcription factor; stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; Tcf20; TCF-20; Transcription factor 20; transcription factor 20 (AR1) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGU0 | |
| TCF20 | |
| Recombinant protein corresponding to Human TCF20. Recombinant protein control fragment (Product #RP-95352). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |