missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEB2 Recombinant Rabbit Monoclonal Antibody (23GB5610)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553912
Description
TCEB2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).Identifiants chimiques
| TCEB2 | |
| 0.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62869, P62870, Q15370 | |
| 67673, 6923, 81807 | |
| A synthesized peptide derived from human TCEB2 (80-118AA). | |
| Antibody |
| 23GB5610 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ELOB | |
| 0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08 | |
| ELOB | |
| Primary | |
| Affinity chromatography |
Spécifications
| TCEB2 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62869, P62870, Q15370 | |
| ELOB | |
| A synthesized peptide derived from human TCEB2 (80-118AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5610 | |
| Unconjugated | |
| ELOB | |
| 0610040H15Rik; 18 kDa, elongin B; cb162; ELOB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; elongin-B; hypothetical protein LOC617277; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII; SIII p18; TCEB 2; Tceb2; tceb2 protein; transcription elongation factor B (SIII) polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2; transcription elongation factor B (SIII), polypeptide 2 (18 kDa, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B); transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); transcription elongation factor B polypeptide 2; transcription elongation factor B polypeptide 2; elongin-B; transcription elongation factor B subunit 2; wu:fb47e08 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 67673, 6923, 81807 | |
| -20°C | |
| Liquid |