missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-I 2000

Numéro de catalogue. 463210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
463210 10 mg
463250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 463210 Fournisseur Tocris Bioscience™ Code fournisseur 4632/10
Il en reste null

TRPM8 blocker

TRPM8 channel blocker. Inhibits icilin -induced TRPM8 channel activation in rTRPM8-expressing CHO cells (IC50 = 53 nM).

Identifiants chimiques

CAS 1159996-20-9
Molecular Formula C23H18F4N2O

Spécifications

Quantity 10 mg
Formula Weight 414.4
Physical Form Solid
Color White
Purity >99%
Molecular Weight (g/mol) 414.404
Chemical Name or Material TC-I 2000
Recommended Storage Store at +4°C