missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBS-TWEEN™ Tablets, MilliporeSigma™
Spécifications
Spécifications
| Physical Form | Tablet |
| Quantity | 1/Ea. |
| Packaging | Plastic Ampoule |
| Synonym | Tris Buffered Saline-TWEEN™ |
| Physical Form | Tablet |
| Quantity | 1/Ea. |
| Packaging | Plastic Ampoule |
| Synonym | Tris Buffered Saline-TWEEN™ |