missing translation for 'onlineSavingsMsg'
Learn More

TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure

Catalog No. AAJ75892K8
Change view
Click to view available options
Quantity:
100 mL
1 L
5 L
Packaging:
Nalgene bottle
Upright carton
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ75892K8 5 L Upright carton
AAJ75892AE 100 mL Nalgene bottle
AAJ75892K2 1 L Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ75892K8 Supplier Thermo Fisher Scientific Supplier No. J75892K8

Used in Western blotting protocols | CAS: 7647-14-5 | ClNa | 58.44 g/mol

Used in Western blotting protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]

Specifications

Concentration 20X
For Use With (Application) Western blotting protocols
pH 7.4
Physical Form Solution
Quantity 5 L
Packaging Upright carton
Grade Molecular Biology, Ultrapure
Chemical Name or Material TBS
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only