missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBK1 Recombinant Rabbit Monoclonal Antibody (4G6C7)

Description
Positive test controls include: Mouse lung, Mouse testis, Rat testis, 293T, HepG2. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MQSTSNHLWL LSDILGQGAT ANVFRGRHKK TGDLFAIKVF NNISFLRPVD VQMREFEVLK KLNHKNIVKL FAIEEETTTR HKVLIMEFCP CGSLYTVLEE.
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling. TBK1 is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-KB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-KB is associated with IKB proteins in the cell cytoplasm, which inhibit NF-KB activity. Phosphorylation of I-KB by IKB kinase (IKK) complex leads to degradation of I-KB and activation of NF-KB. The IKK complex contains IKKa, IKK beta, and IKKK. A novel IKK related kinase was recently identified and designated TBK1 (TANK-binding kinase 1), NAK (NF-KB-activating kinase), and T2K. NAK/TBK1 activates IKK beta through direct phosphorylation. NAK/TBK1 is activated by growth factors and PMA and mediates IKK and NF-kB activation in response to growth factors. NAK/TBK1 functions upstream of NIK and the IKK complex. NAK/TBK1 is also critical in protecting embryonic liver from apoptosis.
Spécifications
Spécifications
| Antigen | TBK1 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 4G6C7 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | TBK1 |
| Gene Accession No. | Q9UHD2, Q9WUN2 |
| Gene Alias | 1200008B05Rik; AI462036; AW048562; FLJ11330; FTDALS4; NAK; NF-kappa-B-activating kinase; NF-kB-activating kinase; serine/threonine protein kinase TBK1; serine/threonine-protein kinase TBK1; T2K; TANK binding kinase 1; TANK-binding kinase 1; tbk; TBK1 |
| Afficher plus de résultats |