missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tazarotene

Catalog No. AC466300010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466300010 1 g
AC466302500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466300010 Supplier Thermo Scientific Chemicals Supplier No. 466300010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118292-40-3
Molecular Formula C21H21NO2S

Specifications

Quantity 1 g
Melting Point 105.0°C
Formula Weight 351.46
InChI Key OGQICQVSFDPSEI-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=C(N=C1)C#CC1=CC=C2SCCC(C)(C)C2=C1
IUPAC Name ethyl 6-[2-(4,4-dimethyl-3,4-dihydro-2H-1-benzothiopyran-6-yl)ethynyl]pyridine-3-carboxylate
Molecular Weight (g/mol) 351.46
Chemical Name or Material Tazarotene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only