accessibility menu, dialog, popup

UserName

CAS RN 118292-40-3

IUPAC Name: ethyl 6-[2-(4,4-dimethyl-3,4-dihydro-2H-1-benzothiopyran-6-yl)ethynyl]pyridine-3-carboxylate
Synonyms: tazarotene tazaroteno tazorac avage tazarotenum tazarotene (product) tazarotene preparation zorac tazarotene (substance)
Molecular Weight (g/mol): 351.46
Molecular Formula: C21H21NO2S
InChi Key: OGQICQVSFDPSEI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(N=C1)C#CC1=CC=C2SCCC(C)(C)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results