missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tauroursodeoxycholic Acid, Sodium Salt, Calbiochem™,
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Packaging | Glass Bottle |
| Percent Purity | ≥97% (TLC) |