missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Acide taurocholique, sel de sodium, Calbiochem™,
Spécifications
Spécifications
| Color | Blanc |
| Quantity | 25 g |
| Packaging | Bouteille en verre |
| Percent Purity | ≥90% (TLC) |