missing translation for 'onlineSavingsMsg'
Learn More

Tartrazine, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013423
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013423 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013423 Supplier Merck Emd Millipore Supplier No. 0332225MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tartrazine is a synthetic azo dye, commonly found in drugs, food products and cosmetics. It can be synthesized by the reaction of sulfanilic acid with 3-carboxy-l-(4-sulfophenyl)-5-pyrazolone.

Specifications

CAS 1934-21-0
Linear Formula C16H9N4 Na3O9S2
MDL Number MFCD00148908
Synonym Acid Yellow 23
Type Analytical Standard
Quantity 25 mg
Formula Weight 534.36
Percent Purity ≥99% (HPLC)
Grade Analytical Standard