missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, 50% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R8376000500A

Quantity 500 mL
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. R8376000500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

87-69-4 , 7732-18-5
150.086
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
CHEBI:15671
C(C(C(=O)O)O)(C(=O)O)O
C4H6O6
FEWJPZIEWOKRBE-JCYAYHJZSA-N
444305
(2R,3R)-2,3-dihydroxybutanedioic acid

Specifications

Tartaric Acid
87-69-4 , 7732-18-5
C4H6O6
FEWJPZIEWOKRBE-JCYAYHJZSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid
444305
500 mL
Natural Poly Bottle
Aqueous Solution
50% (w/v)
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
C(C(C(=O)O)O)(C(=O)O)O
150.086
CHEBI:15671
Odorless
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15°C - 30°C (59°F - 86°F)

SDS
Documents

Documents