missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, 50% (w/v), Ricca Chemical

Catalog No. R8376000120
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R8376000120 120 mL Natural Poly Bottle
R8376000500 500 mL Natural Poly Bottle
R83760001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R8376000120 Supplier Ricca Chemical Company Supplier No. R8376000120A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Appearance Clear colorless to slight yellow liquid
Assay Percent Range 50% (w/v)
Percent Purity 50%
Quantity 120 mL
Odor Odorless
Packaging Natural Poly Bottle
Recommended Storage General