missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ DuraPlex™ 1-Step RT-qPCR Master Mix, no ROX™

Catalog No. A58671
Change view
Click to view available options
Volume:
0.5 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume
A58671 10 mL
A58669 0.5 mL
A58670 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A58671 Supplier Applied Biosystems™ Supplier No. A58671

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Applied Biosystems TaqPath DuraPlex 1-Step RT-qPCR Master Mix is a benchtop-stable, single-tube master mix optimized for rapid, sensitive, and reproducible detection of viral and bacterial pathogens even in the presence of PCR inhibitors.

Applied Biosystems TaqPath DuraPlex 1-Step RT-qPCR Master Mix is a benchtop-stable, single-tube master mix optimized for rapid, sensitive, and reproducible detection of viral and bacterial pathogens even in the presence of PCR inhibitors. This automation-friendly master mix is ideally suited for testing and developing high-throughput molecular diagnostics, as well as for gene expression analysis for high-throughput biologics screening. The 4X concentrated formulation can multiplex six targets in a single well and has intuitive user handling.

TaqPath DuraPlex 1-Step RT-qPCR Master Mix is available in two formulations:

  • With ROX dye as a passive reference (Cat. No.A58666)—recommended for multiplex assays with up to five targets in a single well*
  • Without ROX dye as a passive reference—recommended for multiplex assays with up to six targets in a single well

*Not recommended for use with assay utilizing JUN (or similar emission wavelength) dye.

Benefits of TaqPath DuraPlex 1-Step RT-qPCR Master Mix

  • Enable workflow automation with benchtop-stability of fully assembled reactions for a full workday
  • Optimize your assay for high-order multiplexing of up to six targets in a single reaction
  • Improve handling, in-use stability, and freeze-thaw conditions
  • Increase confidence in detecting both RNA and DNA targets in samples containing challenging PCR inhibitors
  • Facilitate reproducible results with consistent lot-to-lot performance

Support workflow automation

TaqPath DuraPlex 1-Step RT-qPCR Master Mix enables a full workday (over eight hours) of stability of completely assembled reactions (master mix, sample, and assay), helping enable the use of liquid handling and other automation equipment. This fast-cycling master mix can be used effectively for both RNA and DNA detection of bacterial, viral, and fungal targets. The figure below shows consistent sensitivity with a multiplex assay tested at 0 and 8 hours compared to other manufacturers.

Optimized for high-order multiplexing

TaqPath DuraPlex 1-Step RT-qPCR Master Mix has been validated with multiplex assays with up to six targets in a single well with the No ROX formulation and up to five targets in a single well with the ROX formulation. It enables accurate detection of both low and high RNA and DNA concentration samples and provides wide dynamic range. The figure below demonstrates the excellent PCR linearity over an 8-log input range in a 5-plex assay.

Improved handling

TaqPath DuraPlex 1-Step RT-qPCR Master Mix has been formulated to improve in-use handling, including a lower viscosity to allow for more effective pipetting, including with automated liquid-handling platforms. The master mix is stable at room temperature (24°C) for up to 1 week and can be stored under refrigeration (4°C) for up to one month after first use It can be used confidently through eight freeze-thaw cycles without loss of performance. The novel formulation helps ensure that the master mix can be adapted to a variety of unique workflow scenarios. The figure below shows consistent performance of reagents stored for one week at 24°C.

Increase confidence

The unique proprietary TaqPath DuraPlex 1-Step RT-qPCR formulation allows robust performance even in the presence of substances that can inhibit PCR, resulting in effective template detection from both purified RNA and DNA, including challenging clinical sample matrices such as VTM, stool, blood, and urine. The figure below shows performance with common PCR inhibitors compared to master mixes from other manufacturers.

Facilitate reproducibility

This General Purpose Reagent (GPR), labeled 'For Laboratory Use', is manufactured in an ISO 13485-certified and FDA-registered facility that adheres to the principles of current good manufacturing practices (cGMP). All lots of TaqPath DuraPlex 1-Step RT-qPCR Master Mix are functionally tested to help ensure lot-to-lot reproducibility for Ct and dynamic range consistency across a wide variety of assays. The figure below shows consistent performance of three lots of master mix.

Compliance package

In an effort to efficiently support your compliance needs, the following frequently requested documents are made available below.

  • Statement of Quality relating to current good manufacturing practices (cGMP) of TaqPath master mixes
  • Detailed Lot Specific Certificate of Analysis (COA) (see below in Documents & Downloads section)
  • Supplier evaluation support
    • Completed self-assessment
    • ISO certificates by manufacturing site
  • For EU Reach compliance-related inquiries contact LSGChemicalCompliance@thermofisher.com
  • Change Control and Notification Registration

Not finding the compliance document you're looking for? Technical Support is available to help.

Specifications

Content And Storage Store at –15 to –25°C.
Store at room temperature (24°C) for 1 week after first use and 4°C for 1 month after first use.
Detection Method Primer-probe
Form Liquid
Format Single-Tube
GC-Rich PCR Performance Medium
PCR Method 1-step RT-qPCR
Polymerase Applied Biosystems™ AmpliTaq™ DNA Polymerase with Hot Start
Reaction Speed Fast or Standard
For Use With (Equipment) QuantStudio 5 Dx Real‐Time PCR System, QuantStudio 5 Real‐Time PCR System, QuantStudio 7 Pro Dx Real-Time PCR System, QuantStudio 7 Pro Real-Time PCR System, Roche LightCycler PRO System, Bio-Rad CFX Opus 96 Real-Time PCR System
Multiplex Capability Up to 6-plex
Passive Reference Dye None
Product Line TaqPath
Quantity 10 mL
Shipping Condition Dry Ice
Target Specificity RNA only, Both RNA and DNA
For Use With (Application) Microbial Detection, Gene Expression, DNA Quantification, Presence/Absence
Thermostability 8 hr., Assembled Reaction
Hot Start Antibody-based
No. of Reactions 2000 x 20 μL Reactions
Volume 10 mL
Includes dUTP
Inhibitor Tolerance (Simple) High
Show More Show Less