missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Vaginal Microbiota Amplification Control
The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan Vaginal Microbiota assays. This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection.
$550.00
Specifications
| Concentration | 105 Copies/μL |
|---|---|
| Content And Storage | Store at -5 to -30°C. |
| Label or Dye | Unlabeled |
| Product Line | TaqMan |
| Shipping Condition | Gel Packs |
Description
- The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan™ Vaginal Microbiota assays
- It is a multi-target plasmid that includes amplicons for each of the qualified TaqMan Vaginal Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences
- This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection
- The TaqMan Vaginal Microbiota Amplification Control can be included in vaginal microbiota profiling experiments to verify assay performance and to assist with troubleshooting
Specifications
| 105 Copies/μL | |
| Unlabeled | |
| Gel Packs |
| Store at -5 to -30°C. | |
| TaqMan |
For Research Use Only. Not for use in diagnostic procedures.