missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Vaginal Microbiota Amplification Control

Catalog No. 501148729
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501148729 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501148729 Supplier Applied Biosystems™ Supplier No. A32040

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan Vaginal Microbiota assays. This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection.

  • The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan™ Vaginal Microbiota assays
  • It is a multi-target plasmid that includes amplicons for each of the qualified TaqMan Vaginal Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences
  • This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection
  • The TaqMan Vaginal Microbiota Amplification Control can be included in vaginal microbiota profiling experiments to verify assay performance and to assist with troubleshooting

Specifications

Concentration 105 Copies/μL
Content And Storage Store at -5 to -30°C.
Form Liquid
Format Tube(s)
Label or Dye Unlabeled
Product Line TaqMan
Quantity 20 μL
Shipping Condition Gel Packs
Species Non-Human
For Use With (Application) Pathogen Detection
Do you have any positive controls for the vaginal microbiota assays?

The TaqMan Vaginal Microbiota Amplification Control (Cat. No. A32040) is a linearized multi-target plasmid with target sequences for each available vaginal microbiota profiling assay.

Which sample prep kit do you recommend when running vaginal microbiota profiling experiments?

We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for vaginal microbiota profiling experiments.

For Research Use Only. Not for use in diagnostic procedures.