Learn More
Applied Biosystems™ TaqMan™ MicroRNA Reverse Transcription Kit
Provides the necessary components for 200 reactions of optimal performance in TaqMan™ MicroRNA Assays.
Supplier: Applied Biosystems™ 4366596
Description
A highly specific kit that quantitates only mature miRNAs, not precursors. A sensitive kit that conserves limited samples and requires only 1 to 10 nanograms of total RNA. It is fast, simple and scalable 2-step quantitative RT-PCR assay that provides high-quality results in less than three hours.
Components of this kit are used with the RT primer provided with the TaqMan™ MicroRNA Assay to convert miRNA to cDNA.
Order Info
Shipping Conditions: Dry Ice
Specifications
Contains 10X RT buffer, dNTP mix w/dTTp (100M total), RNase inhibitor (20U/μL), and MultiScribe™ RT enzyme (50U/μL) for 200 reactions. Store at -15 to -25°C. | |
High | |
Fast | |
42°C | |
MicroRNA Reverse Transcription Kit | |
MultiScribe™ | |
200 Reactions | |
10X | |
cDNA | |
Separate Components | |
200 bp or less |
Primer-probe | |
2-step RT-qPCR | |
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System | |
TaqMan™ | |
200 reactions | |
Dry Ice | |
miRNA analysis | |
10 X | |
200 Reactions | |
RNA |
Promotions
-
Thermo Fisher Scientific
,
Applied Biosystems
,
Invitrogen
Get Up to $3,000 of qPCR Products with Purchase
Buy up to $3,000 of qPCR products and get a matching amount of qPCR products at no additional cost.Expires: 06/30/25 Redemption Only
Limited Time See Full Promo Details -
Thermo Fisher Scientific
,
Applied Biosystems
,
Invitrogen
Get Up to $3,000 of qPCR Products with Purchase
Buy up to $3,000 of qPCR products and get a matching amount of qPCR products at no additional cost.Expires: 06/30/25
Limited Time See Full Promo Details
For Research Use Only. Not for use in diagnostic procedures.