Learn More
Applied Biosystems™ TaqMan™ MicroRNA Reverse Transcription Kit
Provides the necessary components for 200 reactions of optimal performance in TaqMan™ MicroRNA Assays.
$816.00
Specifications
| Concentration | 10X |
|---|---|
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | 2-step RT-qPCR |
| Reaction Speed | Fast |
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity & Availability | |||||
4366596
|
Applied Biosystems™
4366596 |
200 Reactions |
Each for $816.00
|
|
|||||
Description
A highly specific kit that quantitates only mature miRNAs, not precursors. A sensitive kit that conserves limited samples and requires only 1 to 10 nanograms of total RNA. It is fast, simple and scalable 2-step quantitative RT-PCR assay that provides high-quality results in less than three hours.
Components of this kit are used with the RT primer provided with the TaqMan™ MicroRNA Assay to convert miRNA to cDNA.
Order Info
Shipping Conditions: Dry Ice
Specifications
| 10X | |
| High | |
| Fast | |
| TaqMan | |
| MultiScribe™ | |
| miRNA analysis | |
| cDNA | |
| RNA |
| Primer-probe | |
| 2-step RT-qPCR | |
| 42°C | |
| MicroRNA Reverse Transcription Kit | |
| Dry Ice | |
| 10 X | |
| Separate Components | |
| 200 bp or less |
For Research Use Only. Not for use in diagnostic procedures.