Learn More
Invitrogen™ TaqMan™ Cells-to-CT™ HepatoExpress™ Kit
GREENER_CHOICE
The TaqMan Cells-to-CT HepatoExpress Kit is a complete cell lysate system designed for gene expression analysis directly from hepatocytes without the need of RNA purification.
Supplier:  Invitrogen™ A65789
 View more versions of this product
Catalog No. A65789
Only null left
Description
Description
The TaqMan Cells-to-CT HepatoExpress Kit offers a complete cell lysis and RT-qPCR workflow solution designed for high-throughput gene expression analysis directly from cultured primary hepatocytes without the need for RNA purification. This innovative kit includes HepatoExpress Lysis Solution, HepatoExpress LysisGuard reagent, ezDNase enzyme, SuperScript IV VILO Master Mix, and the TaqMan Fast Advanced Master Mix. The newly developed HepatoExpress Lysis Solution and LysisGuard reagents enable superior lysis performance and sample stability, while the best-in-class RT-qPCR reagents deliver high sensitivity for a seamless end-to-end workflow in approximately 75 minutes.
Features of the TaqMan Cells-to-CT HepatoExpress Kit include:
  • Effortless workflow—simple process that supports high-throughput processing and automation
  • No tedious RNA purification—no columns, beads, heating, or centrifugation
  • Enhanced stability and lysis—efficient primary hepatocyte lysis, while preserving RNA
  • Robust performance—achieve results equivalent to those from purified RNA
  • Maximum flexibility—accommodates extracellular matrix overlays and multiplexing of targets
  • Sustainable solution—REACH-compliant reagents and minimized plastic waste
Optimized chemistry without sacrificing sensitivity
Gene expression analysis from primary hepatocytes is now simplified with our TaqMan Cells-to-CT HepatoExpress Kit. With a strong lysis that penetrates extracellular matrix overlays without degrading RNA, this product enables a rapid workflow without compromising sensitivity. The inclusion of ezDNase enables integrated removal of genomic DNA during cell lysis. After a 10-minute incubation at room temperature, the lysate is ready for reverse transcription using SuperScript IV VILO Master Mix. Once the RT reaction is complete, qPCR analysis is conducted using the TaqMan Fast Advanced qPCR Master Mix and a TaqMan Gene Expression Assay.
Streamlined workflow with proven performance
RNA prepared using the TaqMan Cells-to-CT HepatoExpress technology substantially expedites and streamlines gene expression analysis of primary hepatocytes. Cell lysates exhibit sensitivity and specificity equivalent to RNA extracted using traditional spin column isolation methods.
The lysis procedure simultaneously prepares cell lysates for RT-qPCR and removes genomic DNA in only ten minutes. The lysis step is easily automated with robotic platforms for high-throughput processing of 96-well plates, as it occurs entirely at room temperature. The high-efficiency SuperScript IV VILO RT Master Mix linearly transcribes RNA across a broad input range, even with suboptimal template purity. Additionally, the TaqMan Cells-to-CT HepatoExpress Kit enables multiplexed detection of targets and is compatible with over 700,000 TaqMan Gene Expression Assays.
Specifications
Chemical Identifiers
TaqMan
Kit
2-step RT-qPCR
Yes
Direct Cell Lysis to RT-qPCR Kit
cDNA
Direct Cell Lysis
Hepatocytes, HepaRG, HepG2
Liquid
High
10 min.
ROX
Frozen, Dry Ice
Gene Expression
RNA
Up to 100,000 Cultured Cells
Specifications
HepatoExpress Lysis Solution (Room Temperature), HepatoExpress LysisGuard (-20°C), Express ezDNase (-20°C), SuperScript IV VILO Master Mix (-20°C), SuperScript IV VILO No RT Control (-20°C), TaqMan Fast Advanced Master Mix (4°C). For long-term storage, store HepatoExpress Lysis Buffer at 4°C.
Liquid
High
10 min.
ROX
100 Reactions
cDNA
Direct Cell Lysis
RNA
Up to 100,000 Cultured Cells
TaqMan
Kit
2-step RT-qPCR
Yes
Direct Cell Lysis to RT-qPCR Kit
Frozen, Dry Ice
Gene Expression
100 Reactions
Hepatocytes, HepaRG, HepG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.