missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human IL2 Gene Expression in Activated & Quiescent T-Cells

Catalog No. 4414144
Encompass
Change view
Click to view available options
Quantity:
1 standard plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4414144 1 standard plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4414144 Supplier Applied Biosystems™ Supplier No. 4414144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manual transfer of reagents into plates during routine experimental setups can be time-consuming and tedious. Daily preparation of such plates can negatively impact overall productivity. To ease the stress and minimize the error rate, TaqMan™ Array Plates are designed to reduce redundant pipetting steps and minimize loading errors, making them an ideal solution for fast and easy experimental setup. TaqMan™ Array Plates contain high-quality TaqMan™ Gene Expression Assays (TaqMan™ probe and PCR primer sets) in a convenient, dried-down, 96- or 384-well plate format. Extensive tests have found no difference in performance between TaqMan™ Assays supplied in solution in single tubes and assays supplied dried in a TaqMan™ Array Plate
  • Extensive collections - choose from over 130 predesigned TaqMan™ Gene Signature Arrays and Plates, or customize your own plate, selecting from any of our 1.3 million inventoried assays across 24 species
  • Affordable - minimum order of only one gene signature plate or six custom plates
  • Convenient - dried TaqMan probe and primer sets supplied in 96-well plates; just add master mix and sample, and then run
  • Quick results - Fast format typically provides results in under an hour

Specifications

Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Format 96-well plate
Reaction Speed Standard
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Purification Method Standard
Quantity 1 standard plate
Research Category Cell Cycle & Proliferation, Apoptosis, Biomarker-Related Pathway(s), Oncology, Immunology, Signal Transduction
Species Human
No. of Reactions 1 x 20 μL Reactions
Show More Show Less
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.

For Research Use Only. Not for use in diagnostic procedures.