missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq Pol Bulk (15U/μL)

Catalog No. 11EPTQC200
Change view
Click to view available options
Quantity:
2000U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11EPTQC200 2000U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11EPTQC200 Supplier MP Biomedicals™ Supplier No. 11EPTQC200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

Supplied with standard reaction buffer

Provided with QBiogene 10X Incubation PCR Buffer which is suitable for broad range of reaction conditions.

Specifications

Concentration 15 U/μL
Quantity 2000U
For Use With (Application) Standard PCR
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.